Learn More
GINS3 Mouse anti-Human, Clone: OTI3A12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808798

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a protein subunit of the GINS heterotetrameric complex, which is essential for the initiation of DNA replication and replisome progression in eukaryotes. Alternatively spliced transcript variants encoding distinct isoforms have been described.Specifications
GINS3 | |
Monoclonal | |
Unconjugated | |
GINS3 | |
PSF3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
64785 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3A12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9BRX5 | |
GINS3 | |
Full length human recombit protein of human GINS3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.