Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GINS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100543
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100543 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100543 Supplier Invitrogen™ Supplier No. PA5100543
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GINS3.

This gene encodes a protein subunit of the GINS heterotetrameric complex, which is essential for the initiation of DNA replication and replisome progression in eukaryotes. Alternatively spliced transcript variants encoding distinct isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GINS3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Gins3
Gene Accession No. Q9BRX5, Q9CY94
Gene Alias 2700085M18Rik; AI616142; DNA replication complex GINS protein PSF3; GINS complex subunit 3; GINS complex subunit 3 (Psf3 homolog); Gins3; PSF3; RGD1308153
Gene Symbols Gins3
Host Species Rabbit
Immunogen A synthesized peptide derived from human GINS3(Accession Q9BRX5), corresponding to amino acid residues E130-Q180.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 307639, 64785, 78833
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.