Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GINS4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117144
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117144 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117144 Supplier Invitrogen™ Supplier No. PA5117144
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: MCF7, 293T, HeLa, THP-1, Mouse thymus, Rat fat Immunogen sequence: LEKEKTRPEG EPSSLSPEEL AFAREFMANT ESYLKNVALK HMPPNLQKVD LFRAVPKPDL DSYVFLRVRE RQENILVEPD TDEQRDYVID LEKGSQHLIR.

The yeast heterotetrameric GINS complex is made up of Sld5, Psf1 (GINS1), Psf2 (GINS2), and Psf3 (GINS3). The formation of the GINS complex is essential for the initiation of DNA replication in yeast and Xenopus egg extracts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GINS4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GINS4
Gene Accession No. Q499W2, Q99LZ3, Q9BRT9
Gene Alias 2810037c03rik; 4933405K01Rik; DNA replication complex GINS protein SLD5; DNA replication complex GINS protein SLD5, N-terminally processed; GINS complex subunit 4; GINS complex subunit 4 (Sld5 homolog); Gins4; RGD1307397; SLD5; SLD5 homolog
Gene Symbols GINS4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 108-207 of human GINS4 (NP_1157121).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109145, 290842, 84296
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.