Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GIP Monoclonal Antibody (A2), Invitrogen™
Mouse Monoclonal Antibody
$221.50 - $541.50
Specifications
Antigen | GIP |
---|---|
Clone | C2 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543801
|
Invitrogen™
MA543801 |
20 μL |
Each for $221.50
|
|
|||||
PIMA543800
|
Invitrogen™
MA543800 |
100 μL |
Each for $541.50
|
|
|||||
Description
This gene encodes an incretin hormone and belongs to the glucagon superfamily. The encoded protein is important in maintaining glucose homeostasis as it is a potent stimulator of insulin secretion from pancreatic beta-cells following food ingestion and nutrient absorption. This gene stimulates insulin secretion via its G protein-coupled receptor activation of adenylyl cyclase and other signal transduction pathways. It is a relatively poor inhibitor of gastric acid secretion.Specifications
GIP | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
gastric inhibitory polypeptide; Gip; Glucose-dependent insulinotropic peptide; glucose-dependent insulinotropic polypeptide; Gludins; incretin hormone; Rabbit Anti GIP; RATGLUDINS | |
Gip | |
IgG1 κ | |
Protein A/G | |
Antibody |
C2 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human, Rat | |
P09681 | |
2695 | |
FISD YSIAMDKI conjugated with OVA. | |
Primary | |
-20°C | |
Gip |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title