Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554470
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554470 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554470 Supplier Invitrogen™ Supplier No. PA554470
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KKEGHFSALP SLPVGSHAKV SSPQPRGPRY AEGTFISDYS IAMDKIHQQD FVNWLLAQKG KKND Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 69%.

This gene encodes an incretin hormone and belongs to the glucagon superfamily. The encoded protein is important in maintaining glucose homeostasis as it is a potent stimulator of insulin secretion from pancreatic beta-cells following food ingestion and nutrient absorption. This gene stimulates insulin secretion via its G protein-coupled receptor activation of adenylyl cyclase and other signal transduction pathways. It is a relatively poor inhibitor of gastric acid secretion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gip
Gene Accession No. P09681
Gene Alias gastric inhibitory polypeptide; Gip; Glucose-dependent insulinotropic peptide; glucose-dependent insulinotropic polypeptide; Gludins; incretin hormone; Rabbit Anti GIP; RATGLUDINS
Gene Symbols Gip
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GIP. Recombinant protein control fragment (Product #RP-93344).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2695
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.