Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIPC-2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988663
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8663 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8663 Supplier Bioss Supplier No. BS8586R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GIPC2 (GIPC PDZ Domain Containing Family Member 2) is a Protein Coding gene. An important paralog of this gene is GIPC1.

Specifications

Antigen GIPC-2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Gipc2
Gene Accession No. Q8TF65
Gene Alias 2200002N01Rik; AU021850; GIPC 2; GIPC PDZ domain containing family member 2; GIPC PDZ domain containing family, member 2; GIPC2; PDZ domain protein GIPC2; PDZ domain-containing protein GIPC2; semaF cytoplasmic domain associated protein 2; SemaF cytoplasmic domain-associated protein 2; semaphorin cytoplasmic domain associated protein 2; SEMCAP 2; Semcap2; SEMCAP-2
Gene Symbols Gipc2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GIPC2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54810
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.