Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIPC (GIPC1) Monoclonal Antibody (OTI5C10), OriGene
SDP

Catalog No. 502543871
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3871 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3871 Supplier OriGene Supplier No. CF811961
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GIPC (GIPC1) Monoclonal antibody specifically detects GIPC (GIPC1) in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen GIPC (GIPC1)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5C10
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene CRYGD
Gene Accession No. P07320, P10067
Gene Alias C19orf3; GAIP C-terminus-interacting protein; Gipc; GIPC PDZ domain containing family member 1; GIPC PDZ domain containing family, member 1; Gipc1; GLUT1 C-terminal binding protein; GLUT1 C-terminal-binding protein; GLUT1CBP; Glut1CIP; Hs.6454; IGF-1 receptor interacting protein 1; IIP-1; neurophilin1-IP; NIP; PDZ domain-containing protein GIPC1; regulator of G-protein signaling 19 interacting protein 1; regulator of G-protein signalling 19 interacting protein 1; Rgs19; RGS19-interacting protein 1; RGS19IP1; RGS-GAIP interacting protein C-terminus; RGS-GAIP interacting protein GIPC; RGS-GAIP interacting protein, C-terminus; RGS-GAIP-interacting protein; semaF cytoplasmic domain associated protein; semaF cytoplasmic domain-associated protein 1; SEMCAP; Semcap1; SEMCAP-1; SYNECTIIN; SYNECTIN; Tax interacting protein, clone 2; tax interaction protein 2; TaxIP2; TIP-2
Host Species Mouse
Immunogen Full length human recombinant protein of human CRYGD (NP_008822) produced in HEK293T cell.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1421, 24278
Target Species Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.