Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GIPC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521761
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4. Predicted reactivity: Mouse (100%), Rat (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GIPC1 is a scaffolding protein that regulates cell surface receptor expression and trafficking (Lee et al., 2008).
Specifications
GIPC1 | |
Polyclonal | |
Unconjugated | |
GIPC1 | |
C19orf3; GAIP C-terminus-interacting protein; Gipc; GIPC PDZ domain containing family member 1; GIPC PDZ domain containing family, member 1; Gipc1; GLUT1 C-terminal binding protein; GLUT1 C-terminal-binding protein; GLUT1CBP; Glut1CIP; Hs.6454; IGF-1 receptor interacting protein 1; IIP-1; neurophilin1-IP; NIP; PDZ domain-containing protein GIPC1; regulator of G-protein signaling 19 interacting protein 1; regulator of G-protein signalling 19 interacting protein 1; Rgs19; RGS19-interacting protein 1; RGS19IP1; RGS-GAIP interacting protein C-terminus; RGS-GAIP interacting protein GIPC; RGS-GAIP interacting protein, C-terminus; RGS-GAIP-interacting protein; semaF cytoplasmic domain associated protein; semaF cytoplasmic domain-associated protein 1; SEMCAP; Semcap1; SEMCAP-1; SYNECTIIN; SYNECTIN; Tax interacting protein, clone 2; tax interaction protein 2; TaxIP2; TIP-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10755, 67903 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.47 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O14908, Q9Z0G0 | |
GIPC1 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human GIPC1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction