Learn More
GIPC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5111899
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)KAPPLVE NEEAEPGR Homology: Rat, mouse - 14/15 amino acid residues identical.
GIPC1 is a scaffolding protein that regulates cell surface receptor expression and trafficking (Lee et al., 2008).
Specifications
GIPC1 | |
Polyclonal | |
Unconjugated | |
GIPC1 | |
C19orf3; GAIP C-terminus-interacting protein; Gipc; GIPC1; GLUT1 C-terminal-binding protein; GLUT1CBP; Glut1CIP; Hs.6454; IIP-1; NIP; PDZ domain-containing protein GIPC1; Rgs19; RGS19-interacting protein 1; RGS19IP1; RGS-GAIP-interacting protein; SemaF cytoplasmic domain-associated protein 1; SEMCAP; Semcap1; SEMCAP-1; SYNECTIIN; Synectin; Tax interaction protein 2; TaxIP2; TIP-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10755, 67903, 83823 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
O14908, Q9Z0G0, Q9Z254 | |
GIPC1 | |
Peptide corresponding to amino acid residues 10-24 of human GIPC1 (Intracellular, N-terminus) | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.