Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIPC2 Antibody (1G10), Alexa Fluor™ 594, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP222552AF594

Catalog No. NP2222A94


Only null left
Add to Cart

Description

Description

GIPC2 Monoclonal specifically detects GIPC2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

GIPC2
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
GIPC2
Recombinant human GIPC2 (1-315aa) purified from E. coli
Signal Transduction
54810
Store at 4°C in the dark.
Western Blot, Flow Cytometry, ELISA
1G10
Western Blot, Flow Cytometry, ELISA
FLJ20075, GIPC PDZ domain containing family, member 2, PDZ domain protein GIPC2, PDZ domain-containing protein GIPC2, semaF cytoplasmic domain associated protein 2, semaphorin cytoplasmic domain associated protein 2, SEMCAP2, SEMCAP-2
Mouse
0.1 mL
Primary
Human
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.