Learn More
Invitrogen™ GIPC2 Polyclonal Antibody

Description
Immunogen sequence: MPLKLRGKKK AKSKETAGLV EGEPTGAGGG SLSASRAPAR RLVFHAQLAH GSATGRVEGF SSIQELYAQI AGAFEISPSE ILYCTLNTPK IDMERLLGGQ LGLEDFIFAH VKGIEKEVNV YKSEDSLGLT ITDNGVGYAF IKRIKDGGVI DSVKTICVGD HIESINGENI VGWRHYDVAK KLKELKKEEL FTMKLIEPKK AFEIELRSKA GKSSGEKIGC GRATLRLRSK GPATVEEMPS ETKAKAIEKI DDVLELYMGI RDIDLATTMF EAGKDKVNPD EFAVALDETL GDFAFPDEFV FDVWGVIGDA KRRGL; Positive Samples: U-251MG; Cellular Location: Cytoplasm.
Specifications
Specifications
| Antigen | GIPC2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.16 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Gipc2 |
| Gene Accession No. | Q8TF65 |
| Gene Alias | 2200002N01Rik; AU021850; GIPC 2; GIPC PDZ domain containing family member 2; GIPC PDZ domain containing family, member 2; GIPC2; PDZ domain protein GIPC2; PDZ domain-containing protein GIPC2; semaF cytoplasmic domain associated protein 2; semaF cytoplasmic domain-associated protein 2; semaphorin cytoplasmic domain associated protein 2; SEMCAP 2; SEMCAP2; SEMCAP-2 |
| Gene Symbols | Gipc2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.