Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GIPC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116286
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116286 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116286 Supplier Invitrogen™ Supplier No. PA5116286
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GIPC3 Polyclonal Antibody for Western Blot

Required for postnatal maturation of the hair bundle and long-term survival of hair cells and spiral ganglion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIPC3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GIPC3
Gene Accession No. Q8TF64
Gene Alias C19orf64; DFNB15; DFNB72; DFNB95; DKFZp686J1198; FLJ40925; GIPC PDZ domain containing family member 3; GIPC PDZ domain containing family, member 3; GIPC3; PDZ domain-containing protein GIPC3
Gene Symbols GIPC3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 243-272 amino acids from the C-terminal region of human GIPC3.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126326
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.