Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GIPR Antibody (12C6), Novus Biologicals™

Mouse Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | GIPR |
---|---|
Clone | 12C6 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
GIPR Monoclonal antibody specifically detects GIPR in Mouse, Rat samples. It is validated for ELISASpecifications
GIPR | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Mouse, Rat | |
gastric inhibitory polypeptide receptor, GIP-R, Glucose-dependent insulinotropic polypeptide receptor, MGC126722, PGQTL2 | |
Recombinant Human GIPR protein [UniProt Q0P543] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
12C6 | |
ELISA | |
Unconjugated | |
Mouse | |
GPCR, Neuroscience | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
2696 | |
IgG2a | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title