Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GIPR Antibody (591827) - Azide and BSA Free, R&D Systems™

Mouse Monoclonal Antibody
$181.90 - $487.50
Specifications
Antigen | GIPR |
---|---|
Clone | 591827 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry, Blocking/Neutralizing, CyTOF-ready |
Applications | Flow Cytometry, Blocking Assay, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB82102SP
![]() |
Novus Biologicals
MAB82102SP |
25 μg |
Each for $181.90
|
|
|||||
MAB82102
![]() |
Novus Biologicals
MAB82102 |
100 μg |
Each for $487.50
|
|
|||||
Description
GIPR Monoclonal specifically detects GIPR in Human samples. It is validated for Flow Cytometry, CyTOF-ready, Block/Neutralize.Specifications
GIPR | |
1.0 mg/mL | |
Flow Cytometry, Blocking Assay, CyTOF | |
Unconjugated | |
Mouse | |
GPCR, Neuroscience | |
PBS with No Preservative | |
2696 | |
NS0 mouse myeloma cell line transfected with human GIPR (amino acids 1-466) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
591827 | |
Flow Cytometry, Blocking/Neutralizing, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
gastric inhibitory polypeptide receptor, GIP-R, Glucose-dependent insulinotropic polypeptide receptor, MGC126722, PGQTL2 | |
GIPR | |
IgG1 | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title