Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GIPR Antibody (591827), DyLight 550, R&D Systems™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB82102R
Description
GIPR Monoclonal specifically detects GIPR in Human samples. It is validated for Flow Cytometry.Specifications
GIPR | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GIPR | |
NS0 mouse myeloma cell line transfected with human GIPR (amino acids 1-466) | |
100 μL | |
GPCR, Neuroscience | |
2696 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
591827 | |
Flow Cytometry | |
gastric inhibitory polypeptide receptor, GIP-R, Glucose-dependent insulinotropic polypeptide receptor, MGC126722, PGQTL2 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction