Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GIPR Antibody (591853), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8210PECY55
Description
GIPR Monoclonal antibody specifically detects GIPR in Human samples. It is validated for Flow CytometrySpecifications
GIPR | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
591853 | |
Flow Cytometry | |
gastric inhibitory polypeptide receptor, GIP-R, Glucose-dependent insulinotropic polypeptide receptor, MGC126722, PGQTL2 | |
NS0 mouse myeloma cell line transfected with human GIPR., Met1-Cys466, Accession # P48546 | |
0.1 mL | |
GPCR, Neuroscience | |
2696 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction