Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GIPR Antibody (Gipg013), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP281067AF594
Description
GIPR Monoclonal specifically detects GIPR in Human, Mouse, Rat, Canine samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
GIPR | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
GIPR | |
Human GIPR. | |
0.1 mL | |
GPCR, Neuroscience | |
2696 | |
Human, Mouse, Rat, Canine | |
Purified |
ELISA, Immunofluorescence | |
Gipg013 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
gastric inhibitory polypeptide receptor, GIP-R, Glucose-dependent insulinotropic polypeptide receptor, MGC126722, PGQTL2 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
The original has a Ki value of 7 nM and a Kd value of 6.8 nM for human GIPR. | |
Store at 4°C in the dark. | |
IgG2a λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction