Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GIPR Antibody (Gipg013), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP281067MFV450
Description
GIPR Monoclonal antibody specifically detects GIPR in Human, Mouse, Rat, Canine samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, NeutralizationSpecifications
| GIPR | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| Purified |
| ELISA, Immunocytochemistry, Neutralization | |
| Gipg013 | |
| ELISA, Immunocytochemistry/ Immunofluorescence, Block/Neutralize | |
| gastric inhibitory polypeptide receptor, GIP-R, Glucose-dependent insulinotropic polypeptide receptor, MGC126722, PGQTL2 | |
| Human GIPR. | |
| 0.1 mL | |
| GPCR, Neuroscience | |
| 2696 | |
| Store at 4C in the dark. | |
| IgG2a λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction