Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GIPR, Rabbit anti-Human,Mouse,Rat,Canine, Clone: Gipg013, Novus Biologicals™

Rabbit Monoclonal Antibody
$268.00 - $572.50
Specifications
Antigen | GIPR |
---|---|
Clone | Gipg013 |
Concentration | 1 mg/mL |
Dilution | ELISA, Immunocytochemistry/Immunofluorescence, Block/Neutralize |
Applications | ELISA, Immunofluorescence, Blocking Assay |
Description
GIPR Monoclonal specifically detects GIPR in Human, Mouse, Rat, Canine samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Block/Neutralize.Specifications
GIPR | |
1 mg/mL | |
ELISA, Immunofluorescence, Blocking Assay | |
Unconjugated | |
Rabbit | |
GPCR, Neuroscience | |
PBS | |
2696 | |
Human GIPR. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Gipg013 | |
ELISA, Immunocytochemistry/Immunofluorescence, Block/Neutralize | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Canine | |
gastric inhibitory polypeptide receptor, GIP-R, Glucose-dependent insulinotropic polypeptide receptor, MGC126722, PGQTL2 | |
GIPR | |
IgG λ | |
Protein A purified | |
The original has a Ki value of 7 nM and a Kd value of 6.8 nM for human GIPR. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title