Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIPR Recombinant Human Monoclonal Antibody (32C2), Invitrogen™

Catalog No. FEMA550255
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550255 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550255 Supplier Thermo Scientific Supplier No. MA550255

Human Recombinant Monoclonal Antibody

GIPR encodes a G-protein coupled receptor for gastric inhibitory polypeptide (GIP), which was originally identified as an activity in gut extracts that inhibited gastric acid secretion and gastrin release, but subsequently was demonstrated to stimulate insulin release in the presence of elevated glucose. Mice lacking this gene exhibit higher blood glucose levels with impaired initial insulin response after oral glucose load. Defect in this gene thus may contribute to the pathogenesis of diabetes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIPR
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 32C2
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300, pH 7.4
Gene GIPR
Gene Accession No. P48546
Gene Alias gastric inhibitory peptide receptor, gastric inhibitory polypeptide receptor, GIP receptor, Gippr, Gipr, GIP-R, Glucose-dependent insulinotropic polypeptide receptor, Gm1081, Gm160, PGQTL2, RATGIPPR
Gene Symbols GIPR
Host Species Human
Immunogen Recombinant Macaca fascicularis GIPR protein
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2696
Target Species Human, Non-human Primate
Content And Storage -20°C or -80°C if preferred
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.