Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GIPR Recombinant Mouse Monoclonal Antibody (12C6)
Mouse Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA550096
Description
GIPR Recombinant Monoclonal Antibody for ELISA
GIPR encodes a G-protein coupled receptor for gastric inhibitory polypeptide (GIP), which was originally identified as an activity in gut extracts that inhibited gastric acid secretion and gastrin release, but subsequently was demonstrated to stimulate insulin release in the presence of elevated glucose. Mice lacking this gene exhibit higher blood glucose levels with impaired initial insulin response after oral glucose load. Defect in this gene thus may contribute to the pathogenesis of diabetes.
Specifications
GIPR | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P43219, Q0P543 | |
GIPR | |
Recombinant Mouse Gipr protein. | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG2a |
ELISA, Neutralization | |
12C6 | |
Unconjugated | |
GIPR | |
gastric inhibitory peptide receptor; gastric inhibitory polypeptide receptor; GIP receptor; Gippr; Gipr; GIP-R; Glucose-dependent insulinotropic polypeptide receptor; Gm1081; Gm160; PGQTL2; RATGIPPR | |
Mouse | |
Affinity Chromatography | |
RUO | |
25024, 381853 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction