Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Girdin Antibody (M012), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP323351
Description
Girdin Monoclonal antibody specifically detects Girdin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
Girdin | |
Monoclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
M012 | |
Western Blot 1:250, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 1:50 | |
Akt phosphorylation enhancer, APEGIRDIN, coiled-coil domain containing 88A, Coiled-coil domain-containing protein 88A, G alpha-interacting vesicle-associated protein, G{alpha}-interacting vesicle-associated protein, Girders of actin filament, girdin, GIVFLJ10392, GRDNAKT-phosphorylation enhancer, HKRP1, HkRP1Galpha-interacting vesicle-associated protein, Hook-related protein 1, KIAA1212DKFZp686D0630 | |
Clone (M012) was generated from a recombinant protein that included amino acid residues within the C-terminal region of human Girdin. [UniProt# Q3V6T2] | |
0.1 mL | |
Phospho Specific | |
55704 | |
Store at -20°C. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction