Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Girdin, p Ser1674 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323205
Description
Girdin Polyclonal antibody specifically detects Girdin in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Girdin | |
Polyclonal | |
Western Blot 1:1000 | |
Akt phosphorylation enhancer, APEGIRDIN, coiled-coil domain containing 88A, Coiled-coil domain-containing protein 88A, G alpha-interacting vesicle-associated protein, G{alpha}-interacting vesicle-associated protein, Girders of actin filament, girdin, GIVFLJ10392, GRDNAKT-phosphorylation enhancer, HKRP1, HkRP1Galpha-interacting vesicle-associated protein, Hook-related protein 1, KIAA1212DKFZp686D0630 | |
Phospho-Girdin (Ser-1674) synthetic peptide (coupled to carrier) corresponding to amino acid residues around serine 1674 in human Girdin. This sequence has high homology to rat and mouse Girdin, and has low homology to similar regions in other Girdin family members. [UniProt# Q3V6T2] | |
0.1 mL | |
Phospho Specific | |
55704 | |
Store at -20°C. | |
IgG |
Western Blot | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction