Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GIT1 Antibody (S39B-8), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222423B
Description
GIT1 Monoclonal specifically detects GIT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Knockout Validated.Specifications
GIT1 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
GIT1 | |
Fusion protein amino acids 375-770 (C-terminus) of rat GIT1 | |
0.1 mL | |
28964 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoassay | |
S39B-8 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Knockout Validated | |
ARF GAP GIT1, ARF GTPase-activating protein GIT1, CAT1, CAT-1, Cool-associated and tyrosine-phosphorylated protein 1, G protein-coupled receptor kinase interacting ArfGAP 1, G protein-coupled receptor kinase interactor 1, G protein-coupled receptor kinase-interactor 1, GRK-interacting protein 1 | |
Mouse | |
Protein G purified | |
Primary | |
Detects approx 90kDa. Does not cross-react with GIT2. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction