Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIT1 Monoclonal Antibody (S39B-8), QED Bioscience™
SDP

Catalog No. 502278225
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8225 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8225 Supplier Leinco Technologies Inc Supplier No. 56537100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GIT1 Monoclonal antibody specifically detects GIT1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen GIT1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone S39B-8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene GIT1
Gene Accession No. Q68FF6, Q9Y2X7, Q9Z272
Gene Alias ARF GAP GIT1; ARF GTPase-activating protein GIT1; CAT1; CAT-1; cool-associated and tyrosine-phosphorylated protein 1; G protein-coupled receptor kinase interacting ArfGAP 1; G protein-coupled receptor kinase interactor 1; G protein-coupled receptor kinase-associated ADP ribosylation factor GTPase-activating protein; G protein-coupled receptor kinase-associated ADP ribosylation factor GTPase-activating protein (GIT1); G protein-coupled receptor kinase-interactor 1; GIT ArfGAP 1; Git1; GRK-interacting protein 1; I79_013368; p95Cat
Host Species Mouse
Immunogen Fusion protein corresponding to aa 375-770 (C-terminus) of rat GIT1 (accession no.Q9Z272).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216963, 28964, 83709
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.