Learn More
GIT1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302101A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
GIT1 is a GTPase-activating protein for the ADP ribosylation factor family. It may serve as a scaffold to bring together molecules to form signaling modules controlling vesicle trafficking, adhesion and cytoskeletal organization. It increases the speed of cell migration, as well as the size and rate of formation of protrusions, possibly by targeting PAK1 to adhesions and the leading edge of lamellipodia.Specifications
GIT1 | |
Polyclonal | |
Unconjugated | |
GIT1 | |
ARF GTPase-activating protein GIT1; G protein-coupled receptor kinase-interactor 1; GRK-interacting protein 1; Cool-associated and tyrosine-phosphorylated protein 1; Cat-1; GIT1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
28964 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9Y2X7 | |
GIT1 | |
Between 600 and 650 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.