Learn More
GIT2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | GIT2 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2298
![]() |
Bethyl Laboratories, Inc
IHC00553T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2297
![]() |
Bethyl Laboratories, Inc
IHC00553 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, X. laevis, X. tropicalis, Chicken, Turkey, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda and Orangutan
GTPase-activating protein for the ADP ribosylation factor family.Specifications
GIT2 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
CAT-2, CAT2, ARF GAP GIT2, ARF GTPase-activating protein GIT2, G protein-coupled receptor kinase-interactor 2, GRK-interacting protein 2, cool-associated, tyrosine phosphorylated protein 2, cool-interacting tyrosine-phosphorylated protein 2 | |
GIT2 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q14161 | |
9815 | |
Between 500 and 550 | |
Primary | |
4° C | |
GIT2 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.