Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GITR-Ligand Rat anti-Mouse, Oligo, Clone: MIH44, BD™ AbSeq ™
Rat Monoclonal Antibody
Supplier: BD Biosciences 940428
This item is not returnable.
View return policy
Description
GITR-Ligand Monoclonal antibody specifically detects GITR-Ligand in Mouse samples. It is validated for Single Cell 3' Sequencing
Specifications
GITR-Ligand | |
Monoclonal | |
Oligo | |
Tnfsf18; Gitrl Glucocorticoid-induced-tumor necrosis factor receptor ligand | |
Mouse GITRL Transfected Cell Line | |
RUO | |
Mouse | |
IgG2b κ |
Single Cell 3' Sequencing | |
Clone MIH44 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction