Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GITR Ligand/TNFSF18 Antibody (994529), DyLight 594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB21773M
Description
GITR Ligand/TNFSF18 Monoclonal antibody specifically detects GITR Ligand/TNFSF18 in Mouse samples. It is validated for Flow Cytometry, Neutralization, CyTOFSpecifications
GITR Ligand/TNFSF18 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, Neutralization, CyTOF | |
994529 | |
Flow Cytometry, Neutralization, CyTOF-ready | |
Activation-inducible TNF-related ligand, AITRLMGC138237, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18 | |
Mouse myeloma cell line NS0-derived recombinant mouse GITR Ligand/TNFSF18, Thr47-Ser173, Accession # Q7TS55 | |
0.1 mL | |
Apoptosis | |
8995 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction