Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GITR Ligand/TNFSF18 Antibody (994529), mFluor Violet 500 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB21773MFV500
Description
GITR Ligand/TNFSF18 Monoclonal antibody specifically detects GITR Ligand/TNFSF18 in Mouse samples. It is validated for Flow Cytometry, Neutralization, CyTOFSpecifications
| GITR Ligand/TNFSF18 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Apoptosis | |
| 8995 | |
| Store at 4C in the dark. |
| Flow Cytometry, Neutralization, CyTOF | |
| 994529 | |
| Flow Cytometry, Neutralization, CyTOF-ready | |
| Activation-inducible TNF-related ligand, AITRLMGC138237, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18 | |
| Mouse myeloma cell line NS0-derived recombinant mouse GITR Ligand/TNFSF18, Thr47-Ser173, Accession # Q7TS55 | |
| RUO | |
| Primary | |
| Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction