Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GITR Ligand/TNFSF18 Antibody (YGL386), Alexa Fluor™ 532, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281089AF532
Description
GITR Ligand/TNFSF18 Monoclonal specifically detects GITR Ligand/TNFSF18 in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
GITR Ligand/TNFSF18 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
TNFSF18 | |
Recombinant protein injected into DA rats. | |
0.1 mL | |
Apoptosis | |
8995 | |
Mouse | |
Purified |
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
YGL386 | |
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
Activation-inducible TNF-related ligand, AITRLMGC138237, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
This antibody binds murine GITR Ligand/TNFSF18. | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction