Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GITR Ligand/TNFSF18 Antibody (YGL386), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB281378 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB281378 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB281378 Supplier Novus Biologicals Supplier No. NBP281089MFV610
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GITR Ligand/TNFSF18 Monoclonal antibody specifically detects GITR Ligand/TNFSF18 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen GITR Ligand/TNFSF18
Applications Flow Cytometry
Classification Monoclonal
Clone YGL386
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias Activation-inducible TNF-related ligand, AITRLMGC138237, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18
Host Species Rabbit
Immunogen Recombinant protein injected into DA rats.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 8995
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.