Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GITR/TNFRSF18 Antibody (2375B) [Janelia Fluor™ 669], Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB52412JF669
Description
GITR/TNFRSF18 Monoclonal antibody specifically detects GITR/TNFRSF18 in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunocytochemistry,CyTOFSpecifications
| GITR/TNFRSF18 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse GITR/TNFRSF18, Accession # O35714 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
| 2375B | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Apoptosis | |
| 8784 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction