Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GITR/TNFRSF18 Antibody (CL7787), Novus Biologicals™

Mouse Monoclonal Antibody
$330.00 - $547.00
Specifications
Antigen | GITR/TNFRSF18 |
---|---|
Clone | CL7787 |
Dilution | Western Blot 1 ug/ml |
Applications | Western Blot |
Classification | Monoclonal |
Description
GITR/TNFRSF18 Monoclonal antibody specifically detects GITR/TNFRSF18 in Human samples. It is validated for Western BlotSpecifications
GITR/TNFRSF18 | |
Western Blot 1 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18 | |
This antibody was developed against a Synthetic Peptide of Q9Y5U5. Actual immunogen sequence is proprietary. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
CL7787 | |
Western Blot | |
Unconjugated | |
Mouse | |
Apoptosis | |
PBS, pH 7.2, 40% glycerol | |
8784 | |
IgG2a | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title