Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GITR/TNFRSF18 Antibody (DTA-1), PerCP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP226661PCP
Description
GITR/TNFRSF18 Monoclonal specifically detects GITR/TNFRSF18 in Mouse samples. It is validated for Flow Cytometry.Specifications
GITR/TNFRSF18 | |
Monoclonal | |
PerCP | |
Q9Y5U5 | |
TNFRSF18 | |
Mouse CD25+ CD4+ T cell line NP_683700 | |
0.1 mL | |
Apoptosis | |
8784 | |
Store at 4C in the dark. | |
IgG2b λ |
Flow Cytometry, Immunoprecipitation | |
DTA-1 | |
Flow Cytometry 1 μL/1 million cells | |
Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18 | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction