Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GITR/TNFRSF18 Antibody (OTI9G8), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271388F
Description
GITR/TNFRSF18 Monoclonal specifically detects GITR/TNFRSF18 in Human samples. It is validated for Immunohistochemistry.Specifications
GITR/TNFRSF18 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
TNFRSF18 | |
Recombinant protein fragment corresponding to amino acids 26-162 of human GITR/TNFRSF18 (NP_004186) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG2b |
Immunohistochemistry | |
OTI9G8 | |
Immunohistochemistry | |
Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18 | |
Mouse | |
Affinity Purified | |
Apoptosis | |
8784.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction