Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GITR/TNFRSF18 Antibody (OTI9G8), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271388JF549
Description
GITR/TNFRSF18 Monoclonal specifically detects GITR/TNFRSF18 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| GITR/TNFRSF18 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TNFRSF18 | |
| Recombinant protein fragment corresponding to amino acids 26-162 of human GITR/TNFRSF18 (NP_004186) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI9G8 | |
| Western Blot, Immunohistochemistry | |
| Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis | |
| 8784.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction