Learn More
GITR/TNFRSF18 Antibody (RM0066-7C13), Alexa Fluor™ 350, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NBP212338AF350
Catalog No. NP212338A30
Only null left
Add to Cart
Description
Description
GITR/TNFRSF18 Monoclonal specifically detects GITR/TNFRSF18 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
GITR/TNFRSF18
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
TNFRSF18
Mouse T cells
0.1 mL
8784
Store at 4°C in the dark.
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
RM0066-7C13
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18
Rat
Protein A or G purified
Primary
Mouse
IgG2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.