Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GITR/TNFRSF18 Antibody (T9P4G4*D7), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250337H
Description
GITR/TNFRSF18 Monoclonal specifically detects GITR/TNFRSF18 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
GITR/TNFRSF18 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
TNFRSF18 | |
Peptide Sequence - TEDARSCQFP | |
0.1 mL | |
Apoptosis | |
8784 | |
Store at 4C in the dark. | |
IgG2b κ |
Western Blot, ELISA, Immunohistochemistry | |
T9P4G4*D7 | |
Western Blot, ELISA, Immunohistochemistry | |
Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction