Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GITR/TNFRSF18 Antibody (YGITR765), DyLight 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281090V
Description
GITR/TNFRSF18 Monoclonal specifically detects GITR/TNFRSF18 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
GITR/TNFRSF18 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
TNFRSF18 | |
nB2/6TG cells transfected with mouse GITR/TNFRSF18 injected into DA rats. | |
0.1 mL | |
Apoptosis | |
8784 | |
Mouse | |
Purified |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
YGITR765 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
This antibody binds murine GITR/TNFRSF18. | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction