Learn More
Invitrogen™ GITR (TNFRSF18) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595469
Description
Human TNFRSF18 shares 76.2% amino acid (aa) sequence identity with mouse TNFRSF18. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat kidney tissue, mouse kidney tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Tumor necrosis factor receptor superfamily member 18 (TNFRSF18), also called GITR or AITR is a protein that in humans is encoded by the TNFRSF18 gene. It is mapped to 1p36.33. This gene encodes a member of the TNF-receptor superfamily. The encoded receptor has been shown to have increased expression upon T-cell activation, and it is thought to play a key role in dominant immunological self-tolerance maintained by CD25 (+)CD4 (+) regulatory T cells. Knockout studies in mice also suggest the role of this receptor is in the regulation of CD3-driven T-cell activation and programmed cell death. Three alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
Specifications
GITR (TNFRSF18) | |
Polyclonal | |
Unconjugated | |
Tnfrsf18 | |
Activation-inducible TNFR family receptor; AITR; CD357; GITR; GITR-D; glucocorticoid-induced TNFR-related protein; PRO364; TNF receptor superfamily activation-inducible protein; TNF receptor superfamily member 18; TNFRSF18; Tumor necrosis factor receptor superfamily member 18; tumor necrosis factor receptor superfamily, member 18; UNQ319; UNQ319/PRO364 | |
Rabbit | |
Affinity chromatography | |
RUO | |
21936, 500598, 8784 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
O35714, Q9Y5U5 | |
Tnfrsf18 | |
E.coli-derived human TNFRSF18 recombinant protein (Position: L178-V241). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.