Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GITR/TNFRSF18 Rabbit anti-Human, Clone: 2, Novus Biologicals™

Rabbit Monoclonal Antibody
$342.00 - $524.00
Specifications
| Antigen | GITR/TNFRSF18 |
|---|---|
| Clone | 2 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
Description
GITR/TNFRSF18 Monoclonal specifically detects GITR/TNFRSF18 in Human samples. It is validated for ELISA.Specifications
| GITR/TNFRSF18 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18 | |
| Recombinant Human GITR/TNFRSF18 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 2 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| 0.2 um filtered solution in PBS | |
| 8784 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title