Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GITRL Chimeric Recombinant Rabbit Monoclonal Antibody (YGL386)
GREENER_CHOICE

Catalog No. PIMA548019
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548019 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548019 Supplier Invitrogen™ Supplier No. MA548019
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: This antibody binds murine GITRL.

The tumor necrosis factor (TNF) and TNF receptor (TNFR) gene superfamilies regulate numerous biological functions including cell proliferation, differentiation, and survival through regulating the activation of the transcription factor NF-kappa-B and various mitogen-activated protein kinases. The glucocorticoid-induced tumor necrosis factor receptor (GITR) is an emerging member of this family that is expressed on CD4+ CD25+ regulatory T cells and appears to have crucial immune regulation functions. Its ligand GITRL is expressed in endothelial and antigen-presenting cells and can activate NF-kappa-B, induce both pro- and anti-apoptotic effects, inhibit the suppressive activity of regulatory T cells, and co-stimulate responder T cells through GITR. Dominant negative forms of NIK and TRAF2 expressed in transfected 293 cells substantially inhibited NF-kappa-B activation, suggesting that the GITRL-GITR pathway involves both NIK and TRAF2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GITRL Chimeric
Applications ELISA, Flow Cytometry, Immunocytochemistry
Classification Recombinant Monoclonal
Clone YGL386
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene TNFSF18
Gene Accession No. Q7TS55
Gene Alias Activation-inducible TNF-related ligand; AITR ligand; AITRL; GITR ligand; Gitrl; GITRLigand; Glucocorticoid-induced TNF-related ligand; glucocorticoid-induced TNFR-related protein ligand; glucocorticoid-induced-tumor necrosis factor receptor ligand; hGITRL; RP1-15D23.1; TL6; TNF superfamily member 18; Tnfsf18; TNLG2A; tumor necrosis factor (ligand) superfamily member 18; tumor necrosis factor (ligand) superfamily, member 18; tumor necrosis factor ligand 2A; tumor necrosis factor ligand superfamily member 18; tumor necrosis factor superfamily member 18; UNQ149/PRO175
Gene Symbols TNFSF18
Host Species Rabbit
Immunogen Recombinant protein injected into DA rats.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 240873
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.