Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GITRL Monoclonal Antibody (109114)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA523852
Description
In sandwich ELISAs, no cross-reactivity or interference with recombinant human (rh) APRIL, rhLIGHT, rhTNF-alpha, or rhVEGI is observed. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
The tumor necrosis factor (TNF) and TNF receptor (TNFR) gene superfamilies regulate numerous biological functions including cell proliferation, differentiation, and survival through regulating the activation of the transcription factor NF-kappa-B and various mitogen-activated protein kinases. The glucocorticoid-induced tumor necrosis factor receptor (GITR) is an emerging member of this family that is expressed on CD4+ CD25+ regulatory T cells and appears to have crucial immune regulation functions. Its ligand GITRL is expressed in endothelial and antigen-presenting cells and can activate NF-kappa-B, induce both pro- and anti-apoptotic effects, inhibit the suppressive activity of regulatory T cells, and co-stimulate responder T cells through GITR. Dominant negative forms of NIK and TRAF2 expressed in transfected 293 cells substantially inhibited NF-kappa-B activation, suggesting that the GITRL-GITR pathway involves both NIK and TRAF2.
Specifications
GITRL | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9UNG2 | |
TNFSF18 | |
Sf 21-derived recombinant human GITR Ligand/TNFSF18Glu52-Ser177. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Neutralization | |
109114 | |
Unconjugated | |
TNFSF18 | |
Activation-inducible TNF-related ligand; AITR ligand; AITRL; GITR ligand; Gitrl; GITRLigand; Glucocorticoid-induced TNF-related ligand; glucocorticoid-induced TNFR-related protein ligand; glucocorticoid-induced-tumor necrosis factor receptor ligand; hGITRL; RP1-15D23.1; TL6; TNF superfamily member 18; Tnfsf18; TNLG2A; tumor necrosis factor (ligand) superfamily member 18; tumor necrosis factor (ligand) superfamily, member 18; tumor necrosis factor ligand 2A; tumor necrosis factor ligand superfamily member 18; tumor necrosis factor superfamily member 18; UNQ149/PRO175 | |
Mouse | |
Protein A/G | |
RUO | |
8995 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction