Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Gja10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568749
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568749 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568749 Supplier Invitrogen™ Supplier No. PA568749
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Horse: 86%; Human: 100%; Mouse: 93%; Pig: 91%; Rabbit: 86%; Rat: 86%.

Connexins, such as GJA10, are involved in the formation of gap junctions, intercellular conduits that directly connect the cytoplasms of contacting cells. Each gap junction channel is formed by docking of 2 hemichannels, each of which contains 6 connexin subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gja10
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GJA10
Gene Accession No. 0
Gene Alias connexin 57; connexin 62; connexin-57; Connexin-62; cx57; Cx-57; Cx59; CX62; gap junction alpha-10 protein; gap junction membrane channel protein alpha 10; gap junction protein alpha 10; gap junction protein connexin 57; gap junction protein, alpha 10; gap junction protein, alpha 10, 62kDa; GJA10; LOW QUALITY PROTEIN: gap junction alpha-10 protein; RGD1309630
Gene Symbols GJA10
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of rat Gja10.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 313126
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.