Learn More
Invitrogen™ GJA9 Polyclonal Antibody

Description
This target displays homology in the following species: Dog: 86%; Human: 100%.
Specifications
Specifications
| Antigen | GJA9 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | GJA9 |
| Gene Accession No. | P57773, Q9UKL4 |
| Gene Alias | connexin 36; connexin 59; connexin36; connexin-36; Connexin-58; Connexin-59; Cx36; CX58; Cx59; gap junction alpha 10; Gap junction alpha-10 protein; Gap junction alpha-9 protein; gap junction delta-2 protein; gap junction membrane channel protein alpha 9; gap junction protein alpha 9; gap junction protein delta 2; gap junction protein, alpha 9; gap junction protein, alpha 9, 59kDa; gap junction protein, delta 2; gap junction protein, delta 2, 36kDa; GJA10; GJA9; GJD2 |
| Gene Symbols | GJA9, GJD2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.