Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GJA9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564341
Description
Immunogen sequence: LNGNQLMEKR ETEGKDSKRN YYSRGHRSIP GVAIDGENNM RQSPQTVFSL PANCDWKPRW LRATWGSSTE HENRGSPPKG N Highest antigen sequence identity to the following orthologs: Mouse - 27%, Rat - 26%.
This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
GJA9 | |
Polyclonal | |
Unconjugated | |
GJA9 | |
connexin 36; connexin 59; connexin36; connexin-36; Connexin-58; Connexin-59; Cx36; CX58; Cx59; gap junction alpha 10; Gap junction alpha-10 protein; Gap junction alpha-9 protein; gap junction delta-2 protein; gap junction membrane channel protein alpha 9; gap junction protein alpha 9; gap junction protein delta 2; gap junction protein, alpha 9; gap junction protein, alpha 9, 59kDa; gap junction protein, delta 2; gap junction protein, delta 2, 36kDa; GJA10; GJA9; GJD2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57369, 81025 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P57773, Q9UKL4 | |
GJA9, GJD2 | |
Recombinant protein corresponding to Human Connexin 57. Recombinant protein control fragment (Product #RP-103660). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction