Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gjb1 Rat anti-Mouse, Clone: R5.21C, Abnova™

Catalog No. 89035129 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-129 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-129 Supplier Abnova Corporation Supplier No. MAB15109
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Rat monoclonal antibody raised against native mouse Gjb1.

  • High specificity
  • High reactivity

Specifications

Antigen Gjb1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone R5.21C
Conjugate Unconjugated
Description Rat monoclonal antibody raised against native mouse Gjb1.
Dilution Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Frozen sections) (0.5-1 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS.
Gene Accession No. P28230
Gene Alias AI118175/Cnx32/Cx32/Gjb-1/MGC6217
Gene Symbols Gjb1
Host Species Rat
Immunogen Mouse liver DOC-JR-plasma membranes.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14618
Target Species Mouse
Content And Storage Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.