Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GJC2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159263
Description
GJC2 Polyclonal specifically detects GJC2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GJC2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
connexin46.6, connexin-46.6, Connexin-47, CX46.6, CX47, Gap junction alpha-12 protein, gap junction gamma-2 protein, gap junction protein, alpha 12, 47kDa, gap junction protein, gamma 2, 47kDa, GJA12connexin 47, HLD2, LMPH1C, PMLDAR, SPG44MGC105119 | |
Rabbit | |
47 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:100, Immunohistochemistry-Paraffin | |
Q5T442 | |
GJC2 | |
Synthetic peptides corresponding to GJC2(gap junction protein, gamma 2, 47kDa) The peptide sequence was selected from the middle region of GJC2. Peptide sequence APASRTGSATSAGTVGEQGRPGTHERPGAKPRAGSEKGSASSRDGKTTVW. | |
Affinity purified | |
RUO | |
57165 | |
Human, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction