Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GJC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | GJC2 |
---|---|
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
GJC2 Polyclonal specifically detects GJC2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GJC2 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
57165 | |
IgG | |
Affinity purified |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
connexin46.6, connexin-46.6, Connexin-47, CX46.6, CX47, Gap junction alpha-12 protein, gap junction gamma-2 protein, gap junction protein, alpha 12, 47kDa, gap junction protein, gamma 2, 47kDa, GJA12connexin 47, HLD2, LMPH1C, PMLDAR, SPG44MGC105119 | |
A synthetic peptide of human GJC2 | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title